Learn More
Cayman Chemical Glucagon (hydrochloride), Purity: ≥95%, CAS: 19179-82-9, 5 mg
SDP
Catalog No. 502933346
Encompass_Preferred
A peptide hormone; produced from proglucagon in pancreatic α cells; stimulates glucose production ex vivo in isolated rat hepatocytes; stimulates glucose output from perfused rat livers via increases in gluconeogenesis and glycogenolysis and decreases in glycolysis; increases plasma glucose levels in rats; secretion is suppressed in response to food intake in normal rats and humans but is maintained or elevated in humans with type 2 diabetes
Catalog No. 50-293-3346 Supplier Cayman Chemical Supplier No. 242045 MG
A peptide hormone; produced from proglucagon in pancreatic α cells; stimulates glucose production ex vivo in isolated rat hepatocytes; stimulates glucose output from perfused rat livers via increases in gluconeogenesis and glycogenolysis and decreases in glycolysis; increases plasma glucose levels in rats; secretion is suppressed in response to food intake in normal rats and humans but is maintained or elevated in humans with type 2 diabetes