Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glucan from Black Yeast, TCI America™
SDP

Catalog No. G03311G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G03311G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. G03311G Supplier TCI America Supplier No. G03311G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 9012-72-0
MDL Number MFCD00466924

Specifications

Quantity 1 g
Physical Form Crystalline Powder
Chemical Name or Material Glucan from Black Yeast
EINECSNumber (8)-0098
RTECSNumber LZ3982600
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.