Learn More
Associates of Cape Cod Inc Glucashield, 5.5 mL/vial 5 Pack
SDP
Catalog No. NC0233703
Encompass
Glucashield buffer is used to reconstitute LAL and render the reagent insensitive to (1→3)-ß-D-glucan interference by effectively blocking the Factor G pathway of the endotoxin clotting cascade. For use with Pyrotell lysate (multi-test vials), Pyrotell-T lysate, Pyrochrome kits, and Chromo-LAL lysate.
Catalog No. NC0233703 Supplier Associates of Cape Cod Inc Supplier No. GB0515
Glucashield buffer is used to reconstitute LAL and render the reagent insensitive to (1→3)-ß-D-glucan interference by effectively blocking the Factor G pathway of the endotoxin clotting cascade. For use with Pyrotell lysate (multi-test vials), Pyrotell-T lysate, Pyrochrome kits, and Chromo-LAL lysate.