Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Glucobrassicin potassium | 143231-38-3 | 5 MG
SDP

Catalog No. 5000417570
Encompass_Preferred

Glucobrassicin potassium is a glucosinolate (GSL) isolated from wild carrot (Raphanus raphanistrum L.). It can be used in biological fumigation or allelopathy research for weed management.

  • Isolated from wild carrot (raphanus raphanistrum l.)
  • Can be used in biological fumigation
  • Can be used in allelopathy research for weed management
  • Purity: 95.97%
  • Appearance: solid, pale purple to light pink
  • For research use only

Catalog No. 50-004-17570 Supplier Medchemexpress LLC Supplier No. HYN7659A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Glucobrassicin potassium is a glucosinolate (GSL) isolated from wild carrot (Raphanus raphanistrum L.). It can be used in biological fumigation or allelopathy research for weed management.

  • Isolated from wild carrot (raphanus raphanistrum l.)
  • Can be used in biological fumigation
  • Can be used in allelopathy research for weed management
  • Purity: 95.97%
  • Appearance: solid, pale purple to light pink
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.