Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARKRAY USA Glucocard™ 01 Test Strips

Catalog No. 22355171
Change view
Click to view available options
Quantity:
50 Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-355-171 50 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-355-171 Supplier ARKRAY USA Supplier No. ARK 740050

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Glucocard™ 01 Test Strips for the Glucocard 01 Blood Glucose Meter, an auto-coded meter with a 7 sec test time and 0.3 μL sample size.

  • Sample Size: 0.3 μL
  • Chesmitry: Glucose Oxidase
  • Test Time: 7 Seconds
  • Alternate Site Testing, Dowloadable Capability
  • Open Bottle Shelf Life: 3 Months from Date of Opening
  • For use with The Glucocard 01 Meter Only
  • For in vitro Diagnostic Use Only
  • Critically ill patients should not be tested with the blood glucose meter
  • Not for use with neonates

Specifications

Color Code White
Content And Storage 90 day open shelf life expiration
Clia Complexity Waived
Format Tube
Type Test Strips
Quantity 50 Pk.
Sample Size 0.3 μL
Test Time 7 sec.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.