Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARKRAY USA Glucocard™ Vital™ Test Strips

Catalog No. 22355172
Change view
Click to view available options
Quantity:
50 Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-355-172 50 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-355-172 Supplier ARKRAY USA Supplier No. ARK 760050
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Glucocard™ Vital™ Test Strips for the , small 0.5 uL sample size, glucose oxidase strip chemistry, accurate results in 7 seconds, 250 test memory

  • Sample Size: 0.5 μL
  • Chesmitry: Glucose Oxidase
  • Test Time: 7 Seconds
  • Open Bottle Shelf Life: 3 Months From Date Of Opening
  • For Use With The Glucocard Vital Meter Only
  • For in vitro Diagnostic Use Only
  • Critically ill patients should not be tested with the blood glucose meter
  • Not for use with neonates

Specifications

Color Code White
Content And Storage 90 day open shelf life expiration.
Clia Complexity Waived
Format Tube
Type Test Strips
Quantity 50 Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.