Learn More
Medchemexpress LLC Glucocorticoid receptor agonist-1 | 2166375-82-0 | 99.8% | 569.69 g/mol | C35H39NO6 | 5 MG
SDP
Catalog No. 5000262917
Encompass_Preferred
Glucocorticoid receptor agonist-1 is a research-grade, arylpyrazole-based small molecule that acts as a potent glucocorticoid receptor agonist (reported IC50 = 2.8 nM). It is provided as a purified solid for biochemical and cellular studies.
  • Potent glucocorticoid receptor agonist (IC50 = 2.8 nM).
  • High purity: 99.8%, suitable for analytical and biological assays.
  • Molecular formula C35H39NO6; molecular weight 569.69 g/mol.
  • Available in small research pack sizes, including 5 mg.
  • Provided with datasheet, certificate of analysis, and safety data sheet.
Catalog No. 50-002-62917 Supplier Medchemexpress LLC Supplier No. HY1319745MG
Glucocorticoid receptor agonist-1 is a research-grade, arylpyrazole-based small molecule that acts as a potent glucocorticoid receptor agonist (reported IC50 = 2.8 nM). It is provided as a purified solid for biochemical and cellular studies.
  • Potent glucocorticoid receptor agonist (IC50 = 2.8 nM).
  • High purity: 99.8%, suitable for analytical and biological assays.
  • Molecular formula C35H39NO6; molecular weight 569.69 g/mol.
  • Available in small research pack sizes, including 5 mg.
  • Provided with datasheet, certificate of analysis, and safety data sheet.