Learn More
Cayman Chemical Glucocorticoid Receptor Bioassay, 96 Well
SDP
Catalog No. 502944693
Encompass_Preferred
This GR Assay is an all-inclusive assay system. In addition to the Reporter Cells, included are two optimized media for use during cell culture and in diluting the user's test samples, the reference agonist Dexamethasone, Luciferase Detection Reagent, and a cell cultureready assay plate.
Catalog No. 50-294-4693 Supplier Cayman Chemical Supplier No. 4103396 WELL
This GR Assay is an all-inclusive assay system. In addition to the Reporter Cells, included are two optimized media for use during cell culture and in diluting the user's test samples, the reference agonist Dexamethasone, Luciferase Detection Reagent, and a cell cultureready assay plate.