Learn More
Cayman Chemical Glucocorticoid Resistance Inh
SDP
Supplier:  Cayman Chemical 255535 MG
Encompass_Preferred
An inhibitor of glucocorticoid resistance; inhibits growth of CUTTL1 cells when used in combination with dexamethasone (EC50 = 28 µM) and is less effective when used alone (EC50 = 294 µM); preferentially inhibits growth of CUTTL1 cells expressing high levels of the glucocorticoid receptor over cells expressing low levels; upregulates the gene expression and protein levels of the glucocorticoid receptor; restores glucocorticoid sensitivity to dexamethasone-resistant CUTTL1 cells
Catalog No. 50-301-5695