Learn More
Cayman Chemical β-Glucogallin, Purity: ≥98%, CAS: 13405-60-2, 250 mg
SDP
Catalog No. 502910639
Encompass_Preferred
A plant metabolite and an aldose reductase 2 inhibitor (IC50 = 17 µM); inhibits glucose-induced sorbitol accumulation in lenses isolated from a transgenic mouse model overexpressing AKR1B1; reduces methylglyoxal-induced increases in COX-2, IL-6, IL-8, CCL2, and RAGE levels in ARPE-19 retinal epithelial cells
Catalog No. 50-291-0639 Supplier Cayman Chemical Supplier No. 36010250 MG
A plant metabolite and an aldose reductase 2 inhibitor (IC50 = 17 µM); inhibits glucose-induced sorbitol accumulation in lenses isolated from a transgenic mouse model overexpressing AKR1B1; reduces methylglyoxal-induced increases in COX-2, IL-6, IL-8, CCL2, and RAGE levels in ARPE-19 retinal epithelial cells