Learn More
ATCC Komagataeibacter hansenii
Supplier:  ATCC 10821
Encompass
Komagataeibacter hansenii strain NCIB 1375 is a whole-genome sequenced bacterium that is cited to produce cellulose.
Catalog No. 50-238-4136