Learn More
ATCC Komagataeibacter hansenii; Strain: NQ5
Catalog No. 502383921
Encompass
Komagataeibacter hansenii strain NQ5 was isolated from sugarcane exudate. This bacterial strain produces cellulose.
Catalog No. 50-238-3921 Supplier ATCC Supplier No. 53582
Komagataeibacter hansenii strain NQ5 was isolated from sugarcane exudate. This bacterial strain produces cellulose.