Learn More
Medchemexpress LLC Gluconate Calcium | 299-28-5 | 98.0% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB109210MM/1ML
Encompass_Preferred
Gluconate Calcium is an orally active glucose derivative that demonstrates antioxidant and antiplatelet activation activities. It has been shown to reduce nitric oxide and inflammatory cytokines, inhibit ERK phosphorylation, and exhibit antitumor activity against colorectal cancer. It also improves conditions such as osteoarthritis, intestinal damage, and acute lung injury.
  • Orally active glucose derivative
  • Reduces nitric oxide and inflammatory cytokines
  • Inhibits ERK phosphorylation
  • Possesses antioxidant and antiplatelet activation activities
  • Demonstrates antitumor activity against colorectal cancer
  • Improves osteoarthritis, intestinal damage, and acute lung injury
  • Inhibits thrombin-induced arachidonic acid peroxidation and superoxide anion production in platelets
  • Reduces platelet protein oxidation/nitration, carbonyl group generation, thiol oxidation, and nitrotyrosine formation
Catalog No. 50-004-11816