Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Gluconate (sodium) | 2687959-99-3 | MFCD00064210 | 99.9% | 219.13 g/mol | C5 13CH11NaO7 | 10 MG
SDP

Catalog No. 5000298282
Encompass_Preferred

Gluconate-1-13C sodium is the carbon-1 13C labeled form of sodium D-gluconate supplied as a white to off-white solid, intended for use as an isotopic tracer in metabolic and analytical studies. It is provided in small analytical quantities and has high isotopic purity.

  • Isotopic labeling: carbon-1 13C site-specific label for tracing and NMR studies.
  • High purity: isotopic and chemical purity reported at 99.89% (manufacturer data).
  • Analytical quantities: available in milligram sizes suitable for tracer and analytical work.
  • Physical form: white to off-white solid, easy to dissolve in aqueous media.
  • Characterization data: molecular weight and SMILES provided by manufacturer for analytical confirmation.

Catalog No. 50-002-98282 Supplier Medchemexpress LLC Supplier No. HYB1092AS10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Gluconate-1-13C sodium is the carbon-1 13C labeled form of sodium D-gluconate supplied as a white to off-white solid, intended for use as an isotopic tracer in metabolic and analytical studies. It is provided in small analytical quantities and has high isotopic purity.

  • Isotopic labeling: carbon-1 13C site-specific label for tracing and NMR studies.
  • High purity: isotopic and chemical purity reported at 99.89% (manufacturer data).
  • Analytical quantities: available in milligram sizes suitable for tracer and analytical work.
  • Physical form: white to off-white solid, easy to dissolve in aqueous media.
  • Characterization data: molecular weight and SMILES provided by manufacturer for analytical confirmation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.