Learn More
Medchemexpress LLC Gluconate sodium | 527-07-1 | ≥98.0% | 1 KG
SDP
Supplier:  Medchemexpress LLC HYB1092A1KG
Encompass_Preferred
Gluconate sodium (D-Gluconic acid sodium salt) is an orally active glucose derivative with antioxidant and antiplatelet activation activities.
  • Reduces nitric oxide and inflammatory cytokines (IL-1β and IL-6)
  • Inhibits ERK phosphorylation
  • Exhibits antitumor activity against colorectal cancer
  • Improves osteoarthritis, intestinal damage, and acute lung injury
Catalog No. 50-004-11817