Learn More
Medchemexpress LLC Gluconate sodium | 527-07-1 | MFCD00064210 | ≥98.0% | 1 ML
Supplier: Medchemexpress LLC HYB1092A10MM/1ML
Gluconate sodium (D-Gluconic acid sodium salt) is an orally active glucose derivative with antioxidant and antiplatelet activation activities. It reduces nitric oxide and inflammatory cytokines (IL-1β and IL-6), and inhibits ERK phosphorylation. This compound also exhibits antitumor activity against colorectal cancer and improves various conditions including osteoarthritis, intestinal damage, and acute lung injury.
- Reduces nitric oxide and inflammatory cytokines
- Inhibits ERK phosphorylation
- Possesses antioxidant activity
- Exhibits antiplatelet activation activities
- Shows antitumor activity against colorectal cancer
- Improves osteoarthritis symptoms
- Alleviates intestinal damage
- Protects against acute lung injury