Learn More
Medchemexpress LLC Gluconate sodium (D-Gluconic acid sodium salt) | 527-07-1 | ≥98.0% | 50 G
SDP
Catalog No. 5000413501
Encompass_Preferred
Gluconate sodium (D-Gluconic acid sodium salt) is an orally active glucose derivative. It reduces nitric oxide and inflammatory cytokines (IL-1β and IL-6) and inhibits ERK phosphorylation. It also possesses antioxidant and antiplatelet activation activities, and has demonstrated antitumor activity against colorectal cancer. Furthermore, it has been shown to improve osteoarthritis, intestinal damage, and acute lung injury.
  • Reduces nitric oxide and inflammatory cytokines.
  • Inhibits ERK phosphorylation.
  • Offers antioxidant and antiplatelet activation.
  • Demonstrates antitumor activity against colorectal cancer.
  • Improves osteoarthritis, intestinal damage, and acute lung injury.
  • Regulates hematological parameters.
  • Alleviates intestinal injury.
Catalog No. 50-004-13501 Supplier Medchemexpress LLC Supplier No. HYB1092A50G
Gluconate sodium (D-Gluconic acid sodium salt) is an orally active glucose derivative. It reduces nitric oxide and inflammatory cytokines (IL-1β and IL-6) and inhibits ERK phosphorylation. It also possesses antioxidant and antiplatelet activation activities, and has demonstrated antitumor activity against colorectal cancer. Furthermore, it has been shown to improve osteoarthritis, intestinal damage, and acute lung injury.
  • Reduces nitric oxide and inflammatory cytokines.
  • Inhibits ERK phosphorylation.
  • Offers antioxidant and antiplatelet activation.
  • Demonstrates antitumor activity against colorectal cancer.
  • Improves osteoarthritis, intestinal damage, and acute lung injury.
  • Regulates hematological parameters.
  • Alleviates intestinal injury.