Learn More
Medchemexpress LLC Gluconate sodium (D-gluconic acid sodium salt) | 527-07-1 | ≥98.0% | 500 G
SDP
Supplier:  Medchemexpress LLC HYB1092A500G
Encompass_Preferred
Gluconate sodium (D-Gluconic acid sodium salt) is an orally active glucose derivative. It reduces nitric oxide and inflammatory cytokines, inhibits ERK phosphorylation, and possesses antioxidant and antiplatelet activation activities. Additionally, it demonstrates antitumor activity against colorectal cancer and has shown improvements in osteoarthritis, intestinal damage, and acute lung injury.
  • Reduces nitric oxide and inflammatory cytokines (IL-1β and IL-6)
  • Inhibits ERK phosphorylation
  • Possesses antioxidant and antiplatelet activation activities
  • Exhibits antitumor activity against colorectal cancer
  • Improves osteoarthritis, intestinal damage, and acute lung injury
Catalog No. 50-004-14024