Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gluconic Acid, 50% Soln, MP Biomedicals

Catalog No. ICN211889 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN211889 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN211889 Supplier MP Biomedicals, Inc Supplier No. 0521188925

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 526-95-4
Molecular Formula C6H12O7
Molecular Weight (g/mol) 196.155
InChI Key RGHNJXZEOKUKBD-SQOUGZDYSA-N
Synonym gluconic acid, d-gluconic acid, dextronic acid, maltonic acid, glycogenic acid, glosanto, gluconate, pentahydroxycaproic acid, gluconic acid, d, d-gluconate
PubChem CID 10690
ChEBI CHEBI:33198
IUPAC Name (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoic acid
SMILES C(C(C(C(C(C(=O)O)O)O)O)O)O

Specifications

Formula Weight 196.16
Quantity 100 g
Chemical Name or Material Gluconic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.