Learn More
Medchemexpress LLC HY-N4068 5mg Medchemexpress, Glucoraphanin CAS:21414-41-5 Purity:>98%
SDP
Catalog No. 501933942
Encompass_Preferred
Medchemexpress, HY-N4068 5mg Glucoraphanin CAS:21414-41-5 Glucoraphanin, a natural glucosinolate found in cruciferous vegetable, is a stable precursor of the Nrf2 inducer sulforaphane, which possesses antioxidant, anti-inflammatory, and anti-carcinogenic effects. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3942 Supplier Medchemexpress LLC Supplier No. HYN40685MG
Medchemexpress, HY-N4068 5mg Glucoraphanin CAS:21414-41-5 Glucoraphanin, a natural glucosinolate found in cruciferous vegetable, is a stable precursor of the Nrf2 inducer sulforaphane, which possesses antioxidant, anti-inflammatory, and anti-carcinogenic effects. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.