Learn More
Cayman Chemical GlucosamIn Cholesterol 10mg
SDP
Catalog No. 502916868
Encompass_Preferred
A glucosamine-based lipid conjugate; has been used in the formation of LNPs; forms a stable, ordered bilayer with the glucosamine moiety facing outward to facilitate recognition by GLUT1 on cells; LNPs containing glucosamine cholesterol and encapsulating ceramide induce apoptosis in hypoxic human A549 cells and A549-derived side population cells but not hypoxic mouse L-929 fibroblasts; LNPs containing glucosamine cholesterol and encapsulating paclitaxel and carboplatin reduce tumor volume and increase tumor-associated cytotoxic T cells in an LL/2 murine lung carcinoma model
Catalog No. 50-291-6868 Supplier Cayman Chemical Supplier No. 3944610 MG
A glucosamine-based lipid conjugate; has been used in the formation of LNPs; forms a stable, ordered bilayer with the glucosamine moiety facing outward to facilitate recognition by GLUT1 on cells; LNPs containing glucosamine cholesterol and encapsulating ceramide induce apoptosis in hypoxic human A549 cells and A549-derived side population cells but not hypoxic mouse L-929 fibroblasts; LNPs containing glucosamine cholesterol and encapsulating paclitaxel and carboplatin reduce tumor volume and increase tumor-associated cytotoxic T cells in an LL/2 murine lung carcinoma model