Learn More
Medchemexpress LLC Glucosamine-13C (hydrochloride) | 84247-63-2 | 216.62 | 5 MG
SDP
Catalog No. 5000415302
Encompass_Preferred
Glucosamine-6-13C hydrochloride is a 13C labeled form of Glucosamine hydrochloride, an amino sugar that acts as a significant precursor in the biochemical synthesis of glycosylated proteins and lipids. This product is extensively utilized in research for metabolic studies and cellular pathway analysis, offering precise tracing capabilities due to its isotopic label.
  • Serves as an important precursor in biosynthesis.
  • Utilized in metabolic studies.
  • Aids in cellular pathway analysis.
  • Provides precise tracing capabilities.
Catalog No. 50-004-15302 Supplier Medchemexpress LLC Supplier No. HYN0733S35MG
Glucosamine-6-13C hydrochloride is a 13C labeled form of Glucosamine hydrochloride, an amino sugar that acts as a significant precursor in the biochemical synthesis of glycosylated proteins and lipids. This product is extensively utilized in research for metabolic studies and cellular pathway analysis, offering precise tracing capabilities due to its isotopic label.
  • Serves as an important precursor in biosynthesis.
  • Utilized in metabolic studies.
  • Aids in cellular pathway analysis.
  • Provides precise tracing capabilities.