Learn More
Medchemexpress LLC Glucosamine (D-Glucosamine; Chitosamine) | 3416-24-8 | MFCD01631140 | ≥98.0% | 25 MG
SDP
Catalog No. 5000413924
Encompass_Preferred
Glucosamine (D-Glucosamine) is an amino sugar and a prominent precursor in the biochemical synthesis of glycosylated proteins and lipids, used as a dietary supplement. It is also a natural constituent of glycosaminoglycans in the cartilage matrix and synovial fluid, and when administered exogenously, it exerts pharmacological effects on osteoarthritic cartilage and chondrocytes. It is for research use only and not sold to patients.
  • Amino sugar and precursor in biochemical synthesis.
  • Natural constituent of glycosaminoglycans.
  • Exerts pharmacological effects on osteoarthritic cartilage and chondrocytes.
  • Exhibits dose-dependent DPPH antioxidant activity.
  • Inhibits HIF-1α at protein level.
  • Decreases phosphorylation of p70S6K and S6.
  • Decreases renal expression of α-smooth muscle actin, collagen I, and fibronectin.
Catalog No. 50-004-13924 Supplier Medchemexpress LLC Supplier No. 25MGHYB1125
Glucosamine (D-Glucosamine) is an amino sugar and a prominent precursor in the biochemical synthesis of glycosylated proteins and lipids, used as a dietary supplement. It is also a natural constituent of glycosaminoglycans in the cartilage matrix and synovial fluid, and when administered exogenously, it exerts pharmacological effects on osteoarthritic cartilage and chondrocytes. It is for research use only and not sold to patients.
  • Amino sugar and precursor in biochemical synthesis.
  • Natural constituent of glycosaminoglycans.
  • Exerts pharmacological effects on osteoarthritic cartilage and chondrocytes.
  • Exhibits dose-dependent DPPH antioxidant activity.
  • Inhibits HIF-1α at protein level.
  • Decreases phosphorylation of p70S6K and S6.
  • Decreases renal expression of α-smooth muscle actin, collagen I, and fibronectin.