Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Glucosamine hydrochloride | 66-84-2 | 215.63 | 10 G
SDP

Catalog No. 5000417144
Encompass_Preferred

Glucosamine hydrochloride (D-Glucosamine hydrochloride) is an amino sugar and a prominent precursor in the biochemical synthesis of glycosylated proteins and lipids, used as a dietary supplement. It is also a natural constituent of glycosaminoglycans found in the cartilage matrix and synovial fluid. When administered exogenously, it exhibits pharmacological effects on osteoarthritic cartilage and chondrocytes.

  • Purity: 98.0%
  • Exhibits dose-dependent DPPH antioxidant activity.
  • Short-term treatment inhibits HIF-1α at the protein level, and decreases phosphorylation of p70S6K and S6, which are translation-related proteins.
  • Significantly decreases renal expression of α-smooth muscle actin, collagen I, and fibronectin in obstructed kidneys and TGF-β1-treated renal cells.
  • Appearance: White to off-white solid.

Catalog No. 50-004-17144 Supplier Medchemexpress LLC Supplier No. HYN073310G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Glucosamine hydrochloride (D-Glucosamine hydrochloride) is an amino sugar and a prominent precursor in the biochemical synthesis of glycosylated proteins and lipids, used as a dietary supplement. It is also a natural constituent of glycosaminoglycans found in the cartilage matrix and synovial fluid. When administered exogenously, it exhibits pharmacological effects on osteoarthritic cartilage and chondrocytes.

  • Purity: 98.0%
  • Exhibits dose-dependent DPPH antioxidant activity.
  • Short-term treatment inhibits HIF-1α at the protein level, and decreases phosphorylation of p70S6K and S6, which are translation-related proteins.
  • Significantly decreases renal expression of α-smooth muscle actin, collagen I, and fibronectin in obstructed kidneys and TGF-β1-treated renal cells.
  • Appearance: White to off-white solid.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.