Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Glucosamine (hydrochloride) | 66-84-2 | 215.63 | 25 G
SDP

Catalog No. 5000389528
Encompass_Preferred

Glucosamine hydrochloride is an amino sugar and a prominent precursor in the biochemical synthesis of glycosylated proteins and lipids, used as a dietary supplement. It is also a natural constituent of glycosaminoglycans in the cartilage matrix and synovial fluid, and when administered exogenously, it exerts pharmacological effects on osteoarthritic cartilage and chondrocytes.

  • Exhibits dose-dependent DPPH antioxidant activity.
  • Short-term treatment inhibits HIF-1α at the protein level.
  • Decreases renal expression of α-smooth muscle actin, collagen I, and fibronectin.
  • Purity 98.0%.
  • Appearance is solid.
  • Color is white to off-white.
  • Initial source includes animals, microorganisms, and endogenous metabolite.

Catalog No. 50-003-89528 Supplier Medchemexpress LLC Supplier No. 25GHYN0733
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Glucosamine hydrochloride is an amino sugar and a prominent precursor in the biochemical synthesis of glycosylated proteins and lipids, used as a dietary supplement. It is also a natural constituent of glycosaminoglycans in the cartilage matrix and synovial fluid, and when administered exogenously, it exerts pharmacological effects on osteoarthritic cartilage and chondrocytes.

  • Exhibits dose-dependent DPPH antioxidant activity.
  • Short-term treatment inhibits HIF-1α at the protein level.
  • Decreases renal expression of α-smooth muscle actin, collagen I, and fibronectin.
  • Purity 98.0%.
  • Appearance is solid.
  • Color is white to off-white.
  • Initial source includes animals, microorganisms, and endogenous metabolite.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.