Learn More
Medchemexpress LLC Glucosamine hydrochloride | 66-84-2 | 215.63 | 5 G
SDP
Catalog No. 5000416216
Encompass_Preferred
Glucosamine hydrochloride (D-Glucosamine hydrochloride) is an amino sugar and a prominent precursor in the biochemical synthesis of glycosylated proteins and lipids, used as a dietary supplement. It is a natural constituent of glycosaminoglycans in the cartilage matrix and synovial fluid, and when administered exogenously, it exerts pharmacological effects on osteoarthritic cartilage and chondrocytes.
  • Exhibits dose-dependent DPPH antioxidant activity
  • Inhibits HIF-1α at the protein level and decreases phosphorylation of p70S6K and S6
  • Significantly decreases renal expression of α-smooth muscle actin, collagen I, and fibronectin
Catalog No. 50-004-16216 Supplier Medchemexpress LLC Supplier No. HYN07335G
Glucosamine hydrochloride (D-Glucosamine hydrochloride) is an amino sugar and a prominent precursor in the biochemical synthesis of glycosylated proteins and lipids, used as a dietary supplement. It is a natural constituent of glycosaminoglycans in the cartilage matrix and synovial fluid, and when administered exogenously, it exerts pharmacological effects on osteoarthritic cartilage and chondrocytes.
  • Exhibits dose-dependent DPPH antioxidant activity
  • Inhibits HIF-1α at the protein level and decreases phosphorylation of p70S6K and S6
  • Significantly decreases renal expression of α-smooth muscle actin, collagen I, and fibronectin