Learn More
Medchemexpress LLC Glucosamine hydrochloride | 66-84-2 | MFCD00135831 | 100.0% | 215.63 g/mol | C6H14ClNO5 | 10 MG
SDP
Catalog No. 5000296515
Encompass_Preferred
Glucosamine hydrochloride is the hydrochloride salt of D-(+)-glucosamine, an amino sugar used as an analytical standard and reference material in research. It is commonly applied in method development and quantitative analyses (HPLC, GC, MS), and in studies of glycosylation and cartilage biology.
  • High purity 99.96% for analytical applications.
  • Molecular formula C6H14ClNO5 and molecular weight 215.63 g/mol.
  • Suitable as a reference standard for HPLC, GC, and MS.
  • Commonly used in method development, calibration, and quality control.
  • Supplied in small milligram quantities for analytical workflows.
Catalog No. 50-002-96515 Supplier Medchemexpress LLC Supplier No. HYN0733R10MG
Glucosamine hydrochloride is the hydrochloride salt of D-(+)-glucosamine, an amino sugar used as an analytical standard and reference material in research. It is commonly applied in method development and quantitative analyses (HPLC, GC, MS), and in studies of glycosylation and cartilage biology.
  • High purity 99.96% for analytical applications.
  • Molecular formula C6H14ClNO5 and molecular weight 215.63 g/mol.
  • Suitable as a reference standard for HPLC, GC, and MS.
  • Commonly used in method development, calibration, and quality control.
  • Supplied in small milligram quantities for analytical workflows.