Learn More
Glucosamine Hydrochloride Dietary Supplement, USP, 98-102%, Spectrum™ Chemical
Shop All Spectrum Chemical Mfg. Corp. Products

Description
Specifications
Specifications
| CAS Max % | 100% |
| Ignition Residue | 0.1% |
| Loss on Drying | 1% |
| Packaging | Amber Glass Bottle |
| Quantity | 5 kg |
| Specific Rotation | +70.0° to +73.0 |
| Percent Purity | 98 to 102% |
| Grade | USP Dietary Supplement |
| Physical Form | Crystalline Powder or Lumps |
| Chemical Name or Material | Glucosamine Hydrochloride |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.