Learn More
Apexbio Technology LLC GLUCOSAMINE-SULFATE-20MG
SDP
Catalog No. 5000570800
Encompass_Preferred
Glucosamine sulfate (CAS No 29031-19-4) is a naturally occurring compound primarily used in the biomedical research category of osteoarthritis and joint health Originating from the chitin of shellfish it functions by influencing the synthesis of glycosaminoglycans which are critical for maintaining cartilage structure The mechanism of action involves modulation of inflammatory pathways including inhibition of cartilage-degrading enzymes and the reduction of inflammatory cytokines Common in vitro activity is observed in the low micromolar range reflecting its potency in these biological systems Glucosamine sulfate is frequently applied in cell models and animal studies to assess its therapeutic potential in joint and cartilage health often serving as a candidate in drug screening for anti-inflammatory and chondroprotective agents Experimental concentrations typically depend on the specific research design ensuring optimal relevance and translatability of the findings
Catalog No. 50-005-70800 Supplier Apexbio Technology LLC Supplier No. N273420
Glucosamine sulfate (CAS No 29031-19-4) is a naturally occurring compound primarily used in the biomedical research category of osteoarthritis and joint health Originating from the chitin of shellfish it functions by influencing the synthesis of glycosaminoglycans which are critical for maintaining cartilage structure The mechanism of action involves modulation of inflammatory pathways including inhibition of cartilage-degrading enzymes and the reduction of inflammatory cytokines Common in vitro activity is observed in the low micromolar range reflecting its potency in these biological systems Glucosamine sulfate is frequently applied in cell models and animal studies to assess its therapeutic potential in joint and cartilage health often serving as a candidate in drug screening for anti-inflammatory and chondroprotective agents Experimental concentrations typically depend on the specific research design ensuring optimal relevance and translatability of the findings