Learn More
Medchemexpress LLC Glucosamine (sulfate) | 29031-19-4 | 277.25 | 100 MG
SDP
Catalog No. 5000389878
Encompass_Preferred
Glucosamine sulfate is an amino sugar and a prominent precursor in the biochemical synthesis of glycosylated proteins and lipids, used as a dietary supplement. It is also a natural constituent of glycosaminoglycans in the cartilage matrix and synovial fluid, which, when administered exogenously, exerts pharmacological effects on osteoarthritic cartilage and chondrocytes.
  • Exhibits dose-dependent DPPH antioxidant activity.
  • Inhibits HIF-1α at the protein level with short-term treatment.
  • Decreases phosphorylation of p70S6K and S6, translation-related proteins.
  • Significantly decreases renal expression of α-smooth muscle actin, collagen I, and fibronectin in obstructed kidneys and TGF-β1-treated renal cells.
Catalog No. 50-003-89878 Supplier Medchemexpress LLC Supplier No. HYN0487100MG
Glucosamine sulfate is an amino sugar and a prominent precursor in the biochemical synthesis of glycosylated proteins and lipids, used as a dietary supplement. It is also a natural constituent of glycosaminoglycans in the cartilage matrix and synovial fluid, which, when administered exogenously, exerts pharmacological effects on osteoarthritic cartilage and chondrocytes.
  • Exhibits dose-dependent DPPH antioxidant activity.
  • Inhibits HIF-1α at the protein level with short-term treatment.
  • Decreases phosphorylation of p70S6K and S6, translation-related proteins.
  • Significantly decreases renal expression of α-smooth muscle actin, collagen I, and fibronectin in obstructed kidneys and TGF-β1-treated renal cells.