Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glucosamine Sulfate Potassium Chloride, Dietary Supplement, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18602941 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
12 kg
25 kg
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-602-941 25 kg Poly Bottle
18-602-940 12 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-602-941 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. G129625KGBL

missing translation for 'validMainframeMsg'

G1296, 1296149-08-0, (C6H14NO5)2SO4·2KCl

Spectrum™ Chemical Glucosamine Sulfate Potassium Chloride, Dietary Supplement, USP slows deterioration of cartilage, relieves osteoarthritis (OA) pain and improves joint mobility. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 1296149-08-0

Specifications

CAS Max % 100%
Ignition Residue 26.5-31%
Loss on Drying 1%
Packaging Poly Bottle
Quantity 25 kg
Specific Rotation 47.0° to +53.0
Molecular Weight (g/mol) 422.39
Percent Purity 98 to 102%
Grade Technical
Physical Form Granules
Chemical Name or Material Potassium Ferrocyanide, Trihydrate, Granular
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.