Learn More
Cayman Chemical GlucosdePNdent Insulnotro 10mg
SDP
Catalog No. 502914799
Encompass_Preferred
A pro-atherogenic peptide; induces IκB-α degradation and NF-κB nuclear translocation in macrophage-differentiated THP-1 cells and isolated human aortic endothelial cells at 1 µM; increases atherosclerotic lesion area and plaque formation in ApoE-/- mice
Catalog No. 50-291-4799 Supplier Cayman Chemical Supplier No. 3696810 MG
A pro-atherogenic peptide; induces IκB-α degradation and NF-κB nuclear translocation in macrophage-differentiated THP-1 cells and isolated human aortic endothelial cells at 1 µM; increases atherosclerotic lesion area and plaque formation in ApoE-/- mice