Learn More
Cayman Chemical GlucosdePNdent Insulnotro 10mg
SDP
Catalog No. 502915057
Encompass_Preferred
A peptide fragment of GIP and a GIP receptor antagonist; reduces insulin secretion from BRIN-BD11 cells at 100 nM; increases plasma glucose levels and decreases plasma insulin levels in an ob/ob mouse model of diabetes at 25 nmol/kg
Catalog No. 50-291-5057 Supplier Cayman Chemical Supplier No. 3690810 MG
A peptide fragment of GIP and a GIP receptor antagonist; reduces insulin secretion from BRIN-BD11 cells at 100 nM; increases plasma glucose levels and decreases plasma insulin levels in an ob/ob mouse model of diabetes at 25 nmol/kg