Learn More
Cayman Chemical Glucose-dependent Insulinotropic Polypeptide (1-42) (porcine) (trifluoroacetate salt), Purity: ≥98%, 10 mg
SDP
Catalog No. 502915015
Encompass_Preferred
A peptide incretin hormone; inhibits histamine, pentagastrin, and insulin-induced gastric acid and pepsin secretion, increases glucose-induced insulin release, and stimulates gastric emptying in rats
Catalog No. 50-291-5015 Supplier Cayman Chemical Supplier No. 3690510 MG
A peptide incretin hormone; inhibits histamine, pentagastrin, and insulin-induced gastric acid and pepsin secretion, increases glucose-induced insulin release, and stimulates gastric emptying in rats