Learn More
Cayman Chemical GlucosdePNdent Insulnotro 10mg
Supplier: Cayman Chemical 3686510 MG
An inducer of insulin secretion; enhances glucose-induced insulin release in primary rat pancreatic β-cells at 100 nM to a greater extent than glucose and GIP (1-42)