Learn More
MOLECULAR DEPOT LLC Glucose - 1 mL
Catalog No. 5000013377
Encompass_Preferred
Glucose for Interference Testing is a concentrated glucose solution (250 mg/mL) for spiking into specimens to study the interference of D-glucose in various assays For in vitro research use only
Catalog No. 50-000-13377 Supplier MOLECULAR DEPOT LLC Supplier No. K2010008G1
Glucose for Interference Testing is a concentrated glucose solution (250 mg/mL) for spiking into specimens to study the interference of D-glucose in various assays For in vitro research use only