Learn More
Sigma Aldrich Fine Chemicals Biosciences D-(+)-Glucose powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, >=99.5% | 50-99-7 | MFCD00063774 | 100G
Catalog No. NC0229917
Encompass
D-(+)-Glucose powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, >=99.5% | Purity: >=99.5% | Mol Wt: 180.16 | 50-99-7 | MFCD00063774 | 100G
Catalog No. NC0229917 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. G7021100G
D-(+)-Glucose powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, >=99.5% | Purity: >=99.5% | Mol Wt: 180.16 | 50-99-7 | MFCD00063774 | 100G