Learn More
Medchemexpress LLC Glucose-6-phosphate 10ug
SDP
Catalog No. 5000196083
Encompass_Preferred
Recombinant mouse glucose-6-phosphate isomerase produced in Pichia pastoris and supplied with an N-terminal His tag. The purified enzyme is suitable for biochemical assays, antibody production, and molecular biology applications requiring a native-like GPI protein. Purity is greater than 90% by reducing SDS-PAGE, and microgram-scale sizes are available for small-scale experiments.
  • Expressed in Pichia pastoris for eukaryotic folding and post-translational modifications.
  • N-terminal His tag enables straightforward purification and detection.
  • Purity greater than 90% as determined by reducing SDS-PAGE.
  • Available in microgram quantities for small-scale assays and antibody generation.
  • Suitable for biochemical and immunological studies requiring native enzyme.
Catalog No. 50-001-96083 Supplier Medchemexpress LLC Supplier No. HYP7173810UG
Recombinant mouse glucose-6-phosphate isomerase produced in Pichia pastoris and supplied with an N-terminal His tag. The purified enzyme is suitable for biochemical assays, antibody production, and molecular biology applications requiring a native-like GPI protein. Purity is greater than 90% by reducing SDS-PAGE, and microgram-scale sizes are available for small-scale experiments.
  • Expressed in Pichia pastoris for eukaryotic folding and post-translational modifications.
  • N-terminal His tag enables straightforward purification and detection.
  • Purity greater than 90% as determined by reducing SDS-PAGE.
  • Available in microgram quantities for small-scale assays and antibody generation.
  • Suitable for biochemical and immunological studies requiring native enzyme.