Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Glucose-6-phosphate 10ug
SDP

Supplier:  Medchemexpress LLC HYP7168010UG

Encompass_Preferred

Glucose-6-phosphate isomerase Protein, Human (His) is a recombinant human enzyme that catalyzes the reversible isomerization of glucose-6-phosphate and fructose-6-phosphate. It is provided for use in biochemical assays, enzyme kinetics, antibody validation, and structural studies.

  • Recombinant human enzyme for biochemical assays and structural studies.
  • Microgram-scale supply suitable for antibody validation and small-scale experiments.
  • Approximate molecular weight 65.0 kDa.
  • Contains a His tag to facilitate purification and detection.
  • Store sealed and follow certificate of analysis for specific storage conditions.

Catalog No. 50-001-95811


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.