Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Glucose-6-phosphate 50ug
SDP

Catalog No. 5000196209
Encompass_Preferred

Recombinant mouse glucose-6-phosphate isomerase (GPI) expressed in Pichia pastoris and purified as a His-tagged protein for research applications. The enzyme catalyzes conversion of glucose-6-phosphate to fructose-6-phosphate and is supplied at >90% purity as determined by reducing SDS-PAGE. Available in small-scale quantities suitable for biochemical assays and method development.

  • Recombinant mouse protein expressed in P. pastoris.
  • N-terminal 6xHis tag for easy purification and detection.
  • Purity greater than 90% by reducing SDS-PAGE.
  • Supplied in small-scale sizes suitable for biochemical assays.
  • Intended for research use only.

Catalog No. 50-001-96209 Supplier Medchemexpress LLC Supplier No. HYP7173850UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant mouse glucose-6-phosphate isomerase (GPI) expressed in Pichia pastoris and purified as a His-tagged protein for research applications. The enzyme catalyzes conversion of glucose-6-phosphate to fructose-6-phosphate and is supplied at >90% purity as determined by reducing SDS-PAGE. Available in small-scale quantities suitable for biochemical assays and method development.

  • Recombinant mouse protein expressed in P. pastoris.
  • N-terminal 6xHis tag for easy purification and detection.
  • Purity greater than 90% by reducing SDS-PAGE.
  • Supplied in small-scale sizes suitable for biochemical assays.
  • Intended for research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.