Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Glucose-6-phosphate 5ug
SDP

Catalog No. 5000195520
Encompass_Preferred

Glucose-6-phosphate isomerase Protein, Human (His) is a recombinant human enzyme supplied as a lyophilized powder with a histidine affinity tag, provided for laboratory research and biochemical studies. The dry format supports extended storage and controlled reconstitution for assays.

  • Supplied as lyophilized powder for stable storage.
  • Recombinant human protein source (human).
  • Contains a His affinity tag for purification and detection.
  • Provided for laboratory research applications.

Catalog No. 50-001-95520 Supplier Medchemexpress LLC Supplier No. HYP716805UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Glucose-6-phosphate isomerase Protein, Human (His) is a recombinant human enzyme supplied as a lyophilized powder with a histidine affinity tag, provided for laboratory research and biochemical studies. The dry format supports extended storage and controlled reconstitution for assays.

  • Supplied as lyophilized powder for stable storage.
  • Recombinant human protein source (human).
  • Contains a His affinity tag for purification and detection.
  • Provided for laboratory research applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.