Learn More
Sigma Aldrich Fine Chemicals Biosciences Glucose-6-Phosphate Dehydr
Catalog No. 502901628
Encompass
Glucose-6-Phosphate Dehydrogenase (G6PD) is a ubiquitous enzyme which acts as a catalyst in producing pentose. It also produces NADPH for various biosynthetic and detoxification reactions. Sometimes G6PD provides an alternative to main glycolytic pathway for glucose utilization. It also has its application as DNA markers on X-chromosome.
Catalog No. 50-290-1628 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 10127671001
Glucose-6-Phosphate Dehydrogenase (G6PD) is a ubiquitous enzyme which acts as a catalyst in producing pentose. It also produces NADPH for various biosynthetic and detoxification reactions. Sometimes G6PD provides an alternative to main glycolytic pathway for glucose utilization. It also has its application as DNA markers on X-chromosome.