Learn More
Sigma Aldrich Fine Chemicals Biosciences Glucose 6 Phosphate Dehydr
Catalog No. 502905100
Encompass
Glucose-6-phosphate dehydrogenase (G6P DH) has an active dimeric form whereas its monomeric and tetrameric forms are inactive. At low NADP and NADPH levels and under normal conditions the enzyme exists as equilibrium of tetramers and dimers whereas at high NADP and NADPH levels the equilibrium shifts to inactive tetramers.
Catalog No. 50-290-5100 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 10127655001
Glucose-6-phosphate dehydrogenase (G6P DH) has an active dimeric form whereas its monomeric and tetrameric forms are inactive. At low NADP and NADPH levels and under normal conditions the enzyme exists as equilibrium of tetramers and dimers whereas at high NADP and NADPH levels the equilibrium shifts to inactive tetramers.