Learn More
Sigma Aldrich Fine Chemicals Biosciences Glucose 6 phosphate Dehydr
Supplier:  Sigma Aldrich Fine Chemicals Biosciences G78772KU
Encompass
Glucose 6-phosphate dehydrogenase (G-6-P-DH) is a key regulatory enzyme in the first step of the pentose phosphate pathway. G-6-P-DH is a glycoprotein with a molecular mass of 128 kDa (gel filtration).
Catalog No. 50-290-2279