Learn More
Sigma Aldrich Fine Chemicals Biosciences Glucose 6 phosphate Dehydr
Supplier:  Sigma Aldrich Fine Chemicals Biosciences G576010KU
Encompass
Glucose-6-phosphate Dehydrogenase (G6PD) from Leuconostoc mesenteroides catalyzes the oxidation of glucose 6-phosphate in the presence of nicotinamide adenine dinucleotide (NAD) or nicotinamidenbspadenine dinucleotide phosphate (NADP). It corresponds to a molecular weight of 54 kDa and exists as a homodimer.
Catalog No. 50-290-7130