Learn More
Sigma Aldrich Fine Chemicals Biosciences Glucose 6 phosphate Dehydr
Catalog No. 502914092
Encompass
Glucose 6-phosphate dehydrogenase (G-6-P-DH) is a key regulatory enzyme in the first step of the pentose phosphate pathway. G-6-P-DH is a glycoprotein with a molecular mass of 128 kDa (gel filtration).
Catalog No. 50-291-4092 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. G78775KU
Glucose 6-phosphate dehydrogenase (G-6-P-DH) is a key regulatory enzyme in the first step of the pentose phosphate pathway. G-6-P-DH is a glycoprotein with a molecular mass of 128 kDa (gel filtration).