Learn More
Crescent Chemical Co Inc GLUCOSE-6-PHOSPHATE DEHYDROGEN
SDP
Supplier:  Crescent Chemical Co Inc SE22820.01
Encompass_Preferred
Glucose-6-phosphate dehydrogenase from yeast ca. 140 U/mg protein suspension In 3.2 M ammonium sulfate; pH 6.0.1 mg corresponds to approx. 0.2 ml, 10 mg correspond to approx. 2 ml. Size - 1MG Storage Conditions - +2 °C TO +8 °C Catalog Number - 22820.01 CAS 9001-40-5
Catalog No. 50-247-645