Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Glucose-6-phosphate dehydrogenase | 9001-40-5 | 108.6 kDa | 1KU
SDP

Catalog No. 5000235597
Encompass_Preferred

Glucose-6-phosphate dehydrogenase is the rate-limiting enzyme of the pentose phosphate pathway, provided as a solid enzyme preparation from microorganisms for biochemical research and enzymatic assays involving NADPH generation and oxidative stress pathways.

  • Appearance: White to off-white solid.
  • CAS number: 9001-40-5.
  • Size: 1 KU.
  • Enzyme activity: ≥150 U/mg (solid).
  • Specific activity: ≥500 U/mg (protein).
  • Molecular weight: 108.6 kDa.
  • Optimal pH: 7.0-9.0.
  • Unit definition: One unit oxidizes 1 μmol D-glucose-6-phosphate per minute at pH 7.8 and 37°C.
  • Storage and shipping: Room temperature shipping in continental US; follow certificate of analysis for storage conditions.

Catalog No. 50-002-35597 Supplier Medchemexpress LLC Supplier No. HY1258631KU
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Glucose-6-phosphate dehydrogenase is the rate-limiting enzyme of the pentose phosphate pathway, provided as a solid enzyme preparation from microorganisms for biochemical research and enzymatic assays involving NADPH generation and oxidative stress pathways.

  • Appearance: White to off-white solid.
  • CAS number: 9001-40-5.
  • Size: 1 KU.
  • Enzyme activity: ≥150 U/mg (solid).
  • Specific activity: ≥500 U/mg (protein).
  • Molecular weight: 108.6 kDa.
  • Optimal pH: 7.0-9.0.
  • Unit definition: One unit oxidizes 1 μmol D-glucose-6-phosphate per minute at pH 7.8 and 37°C.
  • Storage and shipping: Room temperature shipping in continental US; follow certificate of analysis for storage conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.