Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Glucose-6-phosphate dehydrogenase | 9001-40-5 | 108.6 kDa | 5KU
SDP

Catalog No. 5000235966
Encompass_Preferred

Glucose-6-phosphate dehydrogenase (G6PD) from a microorganism is the rate-limiting enzyme of the pentose phosphate pathway. Supplied as an enzymatic preparation (5 KU), it generates NADPH for biochemical and enzymatic assays and is intended for research applications including metabolic studies, antioxidant pathway analysis, and disease models such as diabetes, endothelial dysfunction, cancer, and cardiomyopathy. Refer to the product data sheet and certificate of analysis for purity, storage, and handling instructions.

  • Rate-limiting enzyme of the pentose phosphate pathway.
  • Provides NADPH for antioxidant and metabolic assays.
  • Enzyme activity ≥150 U/mg (solid).
  • Specific activity ≥500 U/mg (protein).
  • Unit definition: 1 μmol D-glucose 6-phosphate oxidized per min at pH 7.8, 37°C.
  • Pack size: 5 KU.
  • Source organism: microorganism.
  • Purity and storage details available in data sheet and certificate of analysis.

Catalog No. 50-002-35966 Supplier Medchemexpress LLC Supplier No. HY1258635KU
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Glucose-6-phosphate dehydrogenase (G6PD) from a microorganism is the rate-limiting enzyme of the pentose phosphate pathway. Supplied as an enzymatic preparation (5 KU), it generates NADPH for biochemical and enzymatic assays and is intended for research applications including metabolic studies, antioxidant pathway analysis, and disease models such as diabetes, endothelial dysfunction, cancer, and cardiomyopathy. Refer to the product data sheet and certificate of analysis for purity, storage, and handling instructions.

  • Rate-limiting enzyme of the pentose phosphate pathway.
  • Provides NADPH for antioxidant and metabolic assays.
  • Enzyme activity ≥150 U/mg (solid).
  • Specific activity ≥500 U/mg (protein).
  • Unit definition: 1 μmol D-glucose 6-phosphate oxidized per min at pH 7.8, 37°C.
  • Pack size: 5 KU.
  • Source organism: microorganism.
  • Purity and storage details available in data sheet and certificate of analysis.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.