Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Glucose 6 phosphate dehydrogenase antibody (YA411, rabbit) | 10 UL
SDP

Catalog No. 5000396009
Encompass_Preferred

Glucose 6 phosphate dehydrogenase antibody (YA411) is a rabbit-derived and non-conjugated IgG monoclonal antibody that targets glucose 6 phosphate dehydrogenase.

  • Rabbit-derived and non-conjugated IgG monoclonal antibody
  • Targets glucose 6 phosphate dehydrogenase
  • Applications include metabolism-sugar/lipid metabolism
  • Antibody host is rabbit
  • Target is endogenous metabolite; GLUT
  • Antibody isotype is IgG
  • Antibody type is primary antibody; recombinant
  • Stored at -20°C for 1 year
  • Shipped on blue ice

Catalog No. 50-003-96009 Supplier Medchemexpress LLC Supplier No. HYP8068810UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Glucose 6 phosphate dehydrogenase antibody (YA411) is a rabbit-derived and non-conjugated IgG monoclonal antibody that targets glucose 6 phosphate dehydrogenase.

  • Rabbit-derived and non-conjugated IgG monoclonal antibody
  • Targets glucose 6 phosphate dehydrogenase
  • Applications include metabolism-sugar/lipid metabolism
  • Antibody host is rabbit
  • Target is endogenous metabolite; GLUT
  • Antibody isotype is IgG
  • Antibody type is primary antibody; recombinant
  • Stored at -20°C for 1 year
  • Shipped on blue ice

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.